Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011292.2 | NP_035422.1 | 7 | 596 | ||
GenBank mRNA | AF260271.1 | - | 6 | 566 | ||
AK012331.1 | - | 7 | 566 | |||
AK012445.1 | - | 7 | 566 | |||
AK017388.1 | - | 7 | 566 | |||
AK017427.1 | - | 7 | 566 | |||
AK050296.1 | - | 2 | 2404 | |||
AK053431.1 | - | 3 | 2443 | |||
AK084278.1 | - | 7 | 567 | |||
AK088166.1 | - | 7 | 566 | |||
AK153562.1 | - | 7 | 566 | |||
AK159980.1 | - | 7 | 566 | |||
AK168326.1 | - | 7 | 564 | |||
AK169217.1 | - | 7 | 564 | |||
AK191132.1 | - | 3 | 234 | |||
AK204218.1 | - | 3 | 169 | |||
BC013165.1 | - | 6 | 556 | 2655358 | ||
BC069963.1 | - | 1 | 1243 | |||
BC081435.1 | - | 6 | 542 | 6824107 | ||
BC083166.1 | - | 6 | 547 | 6397429 | ||
BC083329.1 | - | 6 | 545 | 6824346 | ||
BC086937.1 | - | 7 | 563 | 6787671 | ||
BC089319.1 | - | 6 | 545 | 6823996 | ||
BC093526.1 | - | 6 | 545 | 5150493 | ||
U17331.1 | - | 5 | 502 | |||
U17332.1 | - | 5 | 502 |