Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169636.1 | - | 3 | 161 | ||
AV470458.1 | - | 3 | 232 | |||
BC002014.1 | - | 2 | 151 | 3484169 | ||
BC100397.1 | - | 2 | 147 | 6439354 | ||
EU349428.1 | - | 2 | 125 |