Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011300.3 | NP_035430.1 | 6, 7 | 610 | ||
GenBank mRNA | AA221266.1 | - | 1, 2 | 111 | ||
AK135520.1 | - | 6, 7 | 539 | |||
AK160632.1 | - | 6, 7 | 539 | |||
AK166096.1 | - | 6, 7 | 539 | |||
AK169636.1 | - | 6, 7 | 539 | |||
BC002014.1 | - | 5, 6 | 529 | 3484169 | ||
BC086785.1 | - | 515 | ||||
BC099605.1 | - | 5, 6 | 506 | 1528652 | ||
BC100397.1 | - | 5, 6 | 525 | 6439354 | ||
EU349428.1 | - | 5, 6 | 503 |
Gene Symbol: Gm6252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132285.1 | - | 1 | 1039 |