Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011310.2 | NP_035440.1 | 3 | 551 | ||
GenBank mRNA | AF004941.1 | - | 3 | 551 | ||
AK133717.1 | - | 3 | 553 | |||
AK161105.1 | - | 1 | 2298 |