Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011321.3 | NP_035451.1 | 3 | 466 | ||
GenBank mRNA | AK035423.1 | - | 3 | 467 | ||
BC049605.1 | - | 3 | 466 | 6743567 | ||
X06246.1 | - | 3 | 466 |
Gene Symbol: Sbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077421.1 | NP_001070889.1 | 3 | 451 | ||
GenBank mRNA | AK009270.1 | - | 3 | 453 | ||
AK009356.1 | - | 3 | 441 | |||
AK009376.1 | - | 3 | 452 | |||
AK009398.1 | - | 3 | 453 | |||
AK009406.1 | - | 3 | 452 | |||
AK009446.1 | - | 3 | 452 | |||
AK009467.1 | - | 3 | 452 | |||
AK009489.1 | - | 3 | 452 | |||
AK009501.1 | - | 3 | 450 | |||
AK009509.1 | - | 3 | 452 | |||
AK009539.1 | - | 3 | 452 | |||
AK009553.1 | - | 3 | 451 | |||
AK009598.1 | - | 3 | 452 | |||
AK009609.1 | - | 3 | 452 | |||
AK009613.1 | - | 3 | 452 | |||
AK009627.1 | - | 3 | 454 | |||
AK009630.1 | - | 3 | 451 | |||
AK009631.1 | - | 3 | 455 | |||
AK009647.1 | - | 3 | 450 | |||
AK009712.1 | - | 3 | 452 | |||
AK009758.1 | - | 3 | 450 | |||
AK009761.1 | - | 3 | 450 | |||
AK009792.1 | - | 3 | 450 | |||
AK009819.1 | - | 3 | 452 | |||
AK009834.1 | - | 3 | 452 | |||
AK009845.1 | - | 3 | 452 | |||
AK009910.1 | - | 3 | 451 | |||
AK009917.1 | - | 3 | 453 | |||
AK009931.1 | - | 3 | 454 | |||
AK009982.1 | - | 3 | 451 | |||
AK010076.1 | - | 3 | 451 | |||
AK010078.1 | - | 3 | 452 | |||
AK010083.1 | - | 3 | 452 | |||
AK010102.1 | - | 3 | 452 | |||
AK010114.1 | - | 3 | 452 | |||
AK010161.1 | - | 3 | 452 | |||
AK010189.1 | - | 3 | 452 | |||
AK010216.1 | - | 3 | 452 | |||
AK010221.1 | - | 3 | 451 | |||
AK010237.1 | - | 3 | 451 | |||
AK075878.1 | - | 2 | 331 | |||
AK075905.1 | - | 1 | 180 | |||
AK160518.1 | - | 3 | 451 | |||
BC089471.1 | - | 3 | 449 | 30301294 |