Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011331.2 | NP_035461.2 | 2 | 159 | ||
GenBank mRNA | AF065934.1 | - | 2 | 125 | ||
AF065935.1 | - | 2 | 125 | |||
AF065936.1 | - | 2 | 125 | |||
AF065937.1 | - | 2 | 125 | |||
AF065938.1 | - | 2 | 125 | |||
AK012356.1 | - | 2 | 159 | |||
BC027520.1 | - | 2 | 114 | 1548072 | ||
U50712.1 | - | 2 | 158 | |||
U66670.1 | - | 2 | 135 |