Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164059.1 | NP_001157531.1 | 8 | 1222 | ||
NM_011346.2 | NP_035476.1 | 9 | 1330 | |||
GenBank mRNA | AK137509.1 | - | 9 | 1242 | ||
AK157273.1 | - | 9 | 1319 | |||
AK161754.1 | - | 9 | 1322 | |||
AK163066.1 | - | 7 | 787 | |||
AK170753.1 | - | 8 | 1223 | |||
AK172116.1 | - | 9 | 1320 | |||
BC052681.1 | - | 9 | 1314 | 30072021 | ||
M25324.1 | - | 9 | 1322 | |||
M36005.1 | - | 9 | 1268 | |||
M36058.1 | - | 9 | 1268 | |||
X14772.1 | - | 9 | 1271 |