Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011352.2 | NP_035482.1 | 6 | 597 | ||
GenBank mRNA | AB017532.1 | - | 6 | 586 | ||
AF030699.1 | - | 6 | 593 | |||
AF176670.1 | - | 6 | 586 | |||
AK156123.1 | - | 6 | 620 | |||
BC057875.1 | - | 6 | 597 | 5319572 |