Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011355.1 | NP_035485.1 | 2, 3 | 294 | ||
GenBank mRNA | AK142246.1 | - | 2, 3 | 311 | ||
AK153524.1 | - | 2, 3 | 325 | |||
AK154945.1 | - | 2, 3 | 346 | |||
AK155085.1 | - | 2, 3 | 330 | |||
BC003815.1 | - | 2, 3 | 212 | 3600260 | ||
L03215.1 | - | 2, 3 | 191 | |||
M32370.1 | - | 2, 3 | 274 | |||
M38252.1 | - | 2, 3 | 294 | |||
X17463.1 | - | 2, 3 | 982 |