Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081459.2 | NP_001074928.1 | 10 | 2945 | ||
NM_001289538.1 | NP_001276467.1 | 9 | 2845 | |||
NM_001289539.1 | NP_001276468.1 | 10 | 2568 | |||
NM_001289540.1 | NP_001276469.1 | 11 | 2705 | |||
NM_001289541.1 | NP_001276470.1 | 9 | 2521 | |||
NM_001289542.1 | NP_001276471.1 | 9 | 2898 | |||
NM_011363.3 | NP_035493.2 | 10 | 2998 | |||
GenBank mRNA | AF020526.1 | - | 10 | 2639 | ||
AF074329.1 | - | 9 | 2343 | |||
AF380422.1 | - | 9 | 2592 | |||
AF421138.1 | - | 9 | 2539 | |||
AF421139.1 | - | 10 | 2692 | |||
AK148085.1 | - | 11 | 2672 | |||
AK154655.1 | - | 10 | 2879 | |||
AK168439.1 | - | 10 | 2540 | |||
AK170444.1 | - | 10 | 2972 | |||
AK171328.1 | - | 10 | 2954 | |||
AK209185.1 | - | 1 | 180 | |||
AK218299.1 | - | 1 | 206 | |||
BC011422.1 | - | 10 | 2490 | 3156603 | ||
BC051978.1 | - | 9 | 2441 | 6401205 |