Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081459.2 | NP_001074928.1 | 10 | 3355 | ||
NM_001289538.1 | NP_001276467.1 | 9 | 3255 | |||
NM_001289539.1 | NP_001276468.1 | 10 | 2978 | |||
NM_001289540.1 | NP_001276469.1 | 11 | 3115 | |||
NM_001289541.1 | NP_001276470.1 | 9 | 2931 | |||
NM_001289542.1 | NP_001276471.1 | 9 | 3308 | |||
NM_011363.3 | NP_035493.2 | 10 | 3408 | |||
GenBank mRNA | AF074329.1 | - | 9 | 2753 | ||
AI182992.1 | - | 1 | 112 | |||
AK148085.1 | - | 11 | 3082 | |||
AK154655.1 | - | 10 | 3289 | |||
AK168439.1 | - | 10 | 2950 | |||
AK170444.1 | - | 10 | 3382 | |||
AK173145.1 | - | 5 | 1029 |