Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256910.1 | NP_001243839.1 | 2 | 527 | ||
NM_004728.3 | NP_004719.2 | 2 | 408 | |||
XM_011540336.1 | XP_011538638.1 | 2 | 400 | |||
XM_017016910.1 | XP_016872399.1 | 2 | 318 | |||
GenBank mRNA | AK315585.1 | - | 2 | 357 | ||
BC008071.2 | - | 2 | 346 | 2987980 | ||
BX648405.1 | - | 2 | 295 | |||
CR749598.1 | - | 2 | 528 | |||
U41387.1 | - | 2 | 359 |