Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113331.2 | NP_001106802.1 | 2 | 770 | ||
NM_011368.5 | NP_035498.2 | 3 | 372 | |||
GenBank mRNA | AK036980.1 | - | 1 | 2338 | ||
AK049357.1 | - | 3 | 345 | |||
AK155158.1 | - | 2 | 765 | |||
AK203451.1 | - | 2 | 193 | |||
AK211173.1 | - | 2 | 190 | |||
BC036172.1 | - | 3 | 323 | 4988907 | ||
BY341731.1 | - | 2 | 205 | |||
U15784.1 | - | 3 | 262 |