Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011371.2 | NP_035501.2 | 4 | 1051 | ||
GenBank mRNA | AK155119.1 | - | 206 | |||
BC141095.1 | - | 1034 | 9055707 | |||
BC145373.1 | - | 1034 | 9053931 | |||
BC145374.1 | - | 4 | 1001 | 9053932 | ||
Y11274.1 | - | 4 | 1051 |