Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276425.1 | NP_001263354.1 | 6 | 1461 | ||
NM_011373.3 | NP_035503.1 | 5 | 1814 | |||
GenBank mRNA | AJ007310.1 | - | 6 | 1456 | ||
AK007601.1 | - | 6 | 1443 | |||
AK085730.1 | - | 6 | 1452 | |||
AK144884.1 | - | 6 | 1452 | |||
AK153036.1 | - | 6 | 1421 | |||
AK155956.1 | - | 4 | 3955 | |||
AK159712.1 | - | 6 | 1805 | |||
AK161559.1 | - | 6 | 1446 | |||
BC056451.1 | - | 5 | 1267 | 5685866 | ||
BC086784.1 | - | 1402 | 6844232 | |||
Y15779.1 | - | 6 | 1438 | |||
Y15780.1 | - | 5 | 1450 |