Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110350.1 | NP_001103820.1 | 4 | 715 | ||
NM_001110351.1 | NP_001103821.1 | 5 | 592 | |||
NM_011378.2 | NP_035508.2 | 5 | 823 | |||
GenBank mRNA | AK032209.1 | - | 4 | 583 | ||
AK211749.1 | - | 2 | 189 | |||
AK220292.1 | - | 4 | 692 | |||
BC052716.1 | - | 4 | 602 | 6837144 | ||
BC053385.1 | - | 4 | 585 | |||
L38620.2 | - | 3 | 482 | |||
L38621.2 | - | 3 | 482 | |||
U22394.1 | - | 5 | 831 |