Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164480.1 | NP_001157952.1 | 10, 11 | 2571 | ||
NM_001164481.1 | NP_001157953.1 | 11, 12 | 2825 | |||
NM_001164482.1 | NP_001157954.1 | 9, 10 | 2510 | |||
NM_001164568.1 | NP_001158040.1 | 9, 10 | 2540 | |||
NM_011379.4 | NP_035509.4 | 10, 11 | 2794 | |||
GenBank mRNA | AK028151.1 | - | 9, 10 | 1781 | ||
AK137578.1 | - | 10, 11 | 2517 | |||
AK149669.1 | - | 10, 11 | 2517 | |||
AK156280.1 | - | 11, 12 | 2779 | |||
AK169945.1 | - | 9, 10 | 2456 | |||
AK220472.1 | - | 10, 11 | 2707 | |||
BC043679.1 | - | 2, 3 | 931 | |||
BC054824.1 | - | 10, 11 | 2515 | 5324326 | ||
D11374.1 | - | 9, 10 | 2468 |