Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ski | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011385.2 | NP_035515.2 | 5 | 1543 | ||
GenBank mRNA | AF435852.1 | - | 5 | 1522 | ||
AK019148.2 | - | 5 | 600 | |||
BC030841.1 | - | 2 | 320 | |||
BC068305.1 | - | 6 | 1387 | 30093626 | ||
CB722801.1 | - | 2 | 252 |