Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011388.2 | NP_035518.1 | 2 | 527 | ||
GenBank mRNA | AB002693.1 | - | 2 | 527 | ||
AY529655.1 | - | 2 | 527 | |||
AY529656.1 | - | 2 | 527 | |||
BC120703.1 | - | 2 | 508 | 40129626 | ||
BC125290.1 | - | 2 | 508 | 40129805 | ||
D87059.1 | - | 2 | 454 |