Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011390.2 | NP_035520.1 | 2 | 227 | ||
GenBank mRNA | AF087436.1 | - | 2 | 229 | ||
AK052910.1 | - | 2 | 278 | |||
AK083598.1 | - | 2 | 185 | |||
AK131129.1 | - | 2 | 181 | |||
AK143535.1 | - | 2 | 221 | |||
AK149750.1 | - | 2 | 222 | |||
AK166215.1 | - | 2 | 221 | |||
AK169950.1 | - | 2 | 217 | |||
AK171498.1 | - | 2 | 264 | |||
BC051558.1 | - | 2 | 216 | |||
BC053470.1 | - | 2 | 853 | |||
BC059242.1 | - | 2 | 221 | |||
BC060133.1 | - | 2 | 221 | |||
BC079677.1 | - | 2 | 185 | |||
BC141107.1 | - | 2 | 187 | 9055719 |