Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011391.1 | NP_035521.1 | 5 | 781 | ||
GenBank mRNA | AF058054.1 | - | 5 | 781 | ||
AK076612.1 | - | 4 | 603 | |||
BC089606.1 | - | 4 | 549 | |||
BC117803.1 | - | 4 | 545 | 40092297 | ||
BC117804.1 | - | 4 | 545 | 40092299 | ||
BC126960.1 | - | 4 | 545 | 40092496 | ||
CT010191.1 | - | 2 | 344 |