Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011395.2 | NP_035525.1 | 1 | 523 | ||
GenBank mRNA | AF078750.1 | - | 1 | 523 | ||
AF082566.1 | - | 1 | 406 | |||
AK143988.1 | - | 1 | 193 | |||
BC137661.1 | - | 1 | 398 | 8860680 | ||
BC137662.1 | - | 1 | 398 | 8860681 |