Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166450.2 | NP_001159922.1 | 5 | 720 | ||
NM_001290703.1 | NP_001277632.1 | 7 | 909 | |||
NM_001290704.1 | NP_001277633.1 | 7 | 900 | |||
NM_001290705.1 | NP_001277634.1 | 4 | 475 | |||
NM_011398.3 | NP_035528.1 | 5 | 807 | |||
GenBank mRNA | AF076981.1 | - | 5 | 808 | ||
AF155812.1 | - | 4 | 395 | |||
AF155813.1 | - | 4 | 386 | |||
AF535150.1 | - | 7 | 901 | |||
AF535151.1 | - | 7 | 910 | |||
AK148277.1 | - | 5 | 2255 | |||
BC048692.1 | - | 4 | 530 | 6333237 | ||
CX238926.1 | - | 5 | 572 |