Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174078.1 | NP_001167549.1 | 7 | 1466 | ||
NM_001174079.1 | NP_001167550.1 | 7 | 1466 | |||
NM_011417.3 | NP_035547.2 | 7 | 1466 | |||
GenBank mRNA | AK051530.1 | - | 7 | 1255 | ||
AK135308.1 | - | 7 | 1364 | |||
AK141517.1 | - | 4 | 620 | |||
AK142137.1 | - | 7 | 1363 | |||
AK147285.1 | - | 7 | 1420 | |||
AK160825.1 | - | 7 | 1370 | |||
AK166837.1 | - | 7 | 1371 | |||
AK177177.1 | - | 1 | 99 | |||
BC014760.1 | - | 7 | 1277 | |||
BC023830.1 | - | 7 | 1324 | |||
BC032162.1 | - | 7 | 1324 | |||
BC054056.1 | - | 7 | 1308 | |||
BC060229.1 | - | 7 | 1360 | |||
BC061214.1 | - | 7 | 1361 | |||
BC079560.1 | - | 7 | 1466 | 30533489 |