Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252629.1 | NP_001239558.1 | 3 | 270 | ||
NM_011420.2 | NP_035550.1 | 3 | 206 | |||
GenBank mRNA | AK012698.1 | - | 3 | 176 | ||
AK144778.1 | - | 3 | 207 | |||
AK152495.1 | - | 3 | 181 | |||
AK167832.1 | - | 3 | 204 | |||
AY821787.1 | - | 3 | 167 | |||
BC045158.1 | - | 3 | 170 | 6486685 | ||
CJ058980.1 | - | 3 | 207 | |||
EU234021.1 | - | 3 | 270 | |||
U63294.1 | - | 3 | 184 | |||
U77714.1 | - | 3 | 167 | |||
Y12835.1 | - | 3 | 192 |