Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011421.2 | NP_035551.1 | 6 | 1716 | ||
GenBank mRNA | AK088147.1 | - | 6 | 1685 | ||
AK145534.1 | - | 6 | 1685 | |||
AK145702.1 | - | 6 | 1717 | |||
AK148189.1 | - | 6 | 1690 | |||
AK149579.1 | - | 6 | 1685 | |||
AK164167.1 | - | 6 | 1685 | |||
AK164717.1 | - | 6 | 1641 | |||
AK167802.1 | - | 6 | 1640 | |||
AK200204.1 | - | 1 | 36 | |||
AK205297.1 | - | 1 | 36 | |||
BC011304.1 | - | 6 | 1651 | 3964394 | ||
BC034515.1 | - | 6 | 1641 | 4482098 | ||
Z14252.1 | - | 6 | 1662 |