Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291056.1 | NP_001277985.1 | 8 | 1529 | ||
NM_011428.3 | NP_035558.1 | 8 | 1529 | |||
GenBank mRNA | AK078038.1 | - | 8 | 1527 | ||
BC018249.1 | - | 8 | 1417 | 4504644 | ||
M22012.1 | - | 8 | 1473 |