Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011432.2 | NP_035562.1 | 3, 4 | 244 | ||
GenBank mRNA | AK137213.1 | - | 3, 4 | 227 | ||
AK168973.1 | - | 3, 4 | 217 | |||
BC008243.1 | - | 3, 4 | 209 | 2655238 | ||
BC067386.1 | - | 1960 | ||||
BC092266.1 | - | 3, 4 | 209 | 4009300 | ||
U70315.1 | - | 3, 4 | 213 | |||
X96767.1 | - | 3, 4 | 200 |