Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011452.2 | NP_035582.1 | 7 | 1645 | ||
GenBank mRNA | AK005491.1 | - | 7 | 1646 | ||
AK167563.1 | - | 7 | 1521 | |||
AK171850.1 | - | 7 | 1518 | |||
BC064768.1 | - | 7 | 1505 | 30430647 | ||
U96705.2 | - | 7 | 1646 |