Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164524.1 | NP_001157996.1 | 3 | 275 | ||
NM_011453.6 | NP_035583.1 | 3 | 246 | |||
GenBank mRNA | AK014448.1 | - | 3 | 250 | ||
AK017139.1 | - | 3 | 264 | |||
BC051664.1 | - | 3 | 246 | 30018215 | ||
CN704519.1 | - | 3 | 275 |