Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb9d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011460.2 | NP_035590.1 | 7 | 1595 | ||
GenBank mRNA | AK028382.1 | - | 7 | 1596 |