Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011465.4 | NP_035595.2 | 3 | 446 | ||
GenBank mRNA | AF093576.1 | - | 3 | 354 | ||
AK142860.1 | - | 3 | 421 | |||
BC150747.1 | - | 3 | 311 | 9087658 | ||
U87455.1 | - | 3 | 426 |