Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286526.1 | NP_001273455.1 | 5 | 861 | ||
NM_004765.3 | NP_004756.2 | 5 | 861 | |||
XM_011545980.2 | XP_011544282.1 | 5 | 1109 | |||
XM_017023885.1 | XP_016879374.1 | 5 | 1112 | |||
XM_017023886.1 | XP_016879375.1 | 5 | 1117 | |||
XM_017023887.1 | XP_016879376.1 | 5 | 1117 | |||
XM_017023888.1 | XP_016879377.1 | 5 | 1117 | |||
GenBank mRNA | AJ223980.1 | - | 5 | 503 | ||
BC019071.1 | - | 5 | 637 | 4909589 | ||
BC058863.1 | - | 5 | 861 | 5167136 |