Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206804.1 | NP_001193733.1 | 11 | 784 | ||
NM_002757.3 | NP_002748.1 | 11 | 1368 | |||
NM_145160.2 | NP_660143.1 | 11 | 1368 | |||
XM_011521787.2 | XP_011520089.1 | 11 | 1445 | |||
XM_011521788.2 | XP_011520090.1 | 11 | 1445 | |||
XM_017022414.1 | XP_016877903.1 | 11 | 1445 | |||
GenBank mRNA | AK057521.1 | - | 5 | 496 | ||
AK293459.1 | - | 11 | 784 | |||
AK315435.1 | - | 11 | 915 | |||
BC008838.2 | - | 11 | 1025 | 4100679 | ||
BT006780.1 | - | 11 | 715 | |||
CR542229.1 | - | 11 | 715 | |||
U71087.1 | - | 11 | 776 | |||
U71088.2 | - | 11 | 720 |