Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011503.4 | NP_035633.1 | 12, 13 | 1033 | ||
GenBank mRNA | AF263345.1 | - | 12, 13 | 1040 | ||
AK088364.1 | - | 12, 13 | 1055 | |||
AK133419.1 | - | 12, 13 | 1034 | |||
AK211462.1 | - | 4, 5 | 283 | |||
BC003477.1 | - | 12, 13 | 1023 | |||
BC008102.1 | - | 12, 13 | 1043 | 3966319 | ||
D42068.1 | - | 12, 13 | 1025 | |||
U19520.1 | - | 12, 13 | 1047 |