Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvalb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013645.3 | NP_038673.2 | 4 | 848 | ||
GenBank mRNA | BC027424.1 | - | 4 | 848 | 4925213 | |
X67141.1 | - | 1 | 522 |