Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281880.1 | NP_001268809.1 | 10 | 1295 | ||
NM_013660.4 | NP_038688.2 | 9 | 1231 | |||
GenBank mRNA | AK088653.1 | - | 6 | 513 | ||
AK132688.1 | - | 10 | 1221 | |||
AK150911.1 | - | 8 | 1123 | |||
BC049780.1 | - | 9 | 1128 | 6509473 | ||
U69535.1 | - | 7 | 758 |