Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 10 | 1126 | ||
NM_001654.4 | NP_001645.1 | 10 | 1117 | |||
XM_006724529.2 | XP_006724592.1 | 10 | 1053 | |||
XM_011543906.2 | XP_011542208.1 | 10 | 2443 | |||
XM_011543908.2 | XP_011542210.1 | 10 | 2427 | |||
XM_011543909.2 | XP_011542211.1 | 5 | 418 | |||
GenBank mRNA | AB208831.1 | - | 9 | 2302 | ||
AK130043.1 | - | 10 | 1010 | |||
AK297453.1 | - | 8 | 840 | |||
AK300975.1 | - | 10 | 973 | |||
BC002466.2 | - | 10 | 997 | 3347509 | ||
BC007514.2 | - | 10 | 1004 | 2966604 | ||
BT019864.1 | - | 9 | 906 | |||
KU177914.1 | - | 9 | 906 | |||
KU177915.1 | - | 9 | 921 | |||
M13829.1 | - | 1 | 33 | |||
X04790.1 | - | 10 | 1100 |