Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130456.1 | NP_001123928.1 | 8 | 833 | ||
NM_013662.2 | NP_038690.1 | 8 | 944 | |||
GenBank mRNA | AF036585.1 | - | 7 | 700 | ||
AK139358.1 | - | 8 | 753 | |||
AK143229.1 | - | 8 | 834 | |||
AK166077.1 | - | 8 | 936 | |||
BC139415.1 | - | 7 | 699 | 8862437 |