Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277982.1 | NP_001264911.1 | 6 | 585 | ||
NM_001277983.1 | NP_001264912.1 | 6 | 697 | |||
NM_001277985.1 | NP_001264914.1 | 6 | 697 | |||
NM_001277986.1 | NP_001264915.1 | 6 | 697 | |||
NM_013669.2 | NP_038697.1 | 6 | 697 | |||
GenBank mRNA | AK051176.1 | - | 6 | 692 | ||
AK147087.1 | - | 6 | 688 | |||
AK159570.1 | - | 6 | 663 | |||
BC031773.1 | - | 6 | 585 | 4511467 | ||
BC052454.1 | - | 6 | 697 | 6410678 | ||
M83985.1 | - | 6 | 622 |