Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276479.1 | NP_001263408.1 | 3 | 313 | ||
NM_001276480.1 | NP_001263409.1 | 2 | 277 | |||
NM_004793.3 | NP_004784.2 | 2 | 598 | |||
NR_076392.1 | 3 | 296 | ||||
XM_011528441.2 | XP_011526743.1 | 2 | 641 | |||
GenBank mRNA | AK056366.1 | - | 2 | 277 | ||
AK096433.1 | - | 3 | 326 | |||
AK096626.1 | - | 3 | 313 | |||
AK127867.1 | - | 3 | 378 | |||
AK298531.1 | - | 3 | 277 | |||
AK309720.1 | - | 3 | 418 | |||
AL096720.1 | - | 2 | 144 | |||
BC000235.1 | - | 2 | 477 | 3350958 | ||
BC109218.1 | - | 2 | 188 | |||
BI906625.1 | - | 2 | 629 | |||
U02389.1 | - | 2 | 574 |