Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277982.1 | NP_001264911.1 | 29 | 3374 | ||
NM_001277983.1 | NP_001264912.1 | 26 | 3396 | |||
NM_001277985.1 | NP_001264914.1 | 20 | 2603 | |||
NM_001277986.1 | NP_001264915.1 | 30 | 3570 | |||
NM_013669.2 | NP_038697.1 | 30 | 3585 | |||
GenBank mRNA | AB093255.1 | - | 18 | 4623 | ||
AK139322.1 | - | 1 | 90 | |||
AK147087.1 | - | 26 | 3387 | |||
AK159570.1 | - | 20 | 2569 | |||
AK161905.1 | - | 2 | 725 | |||
BC052454.1 | - | 31 | 3741 | 6410678 | ||
M83985.1 | - | 30 | 3509 |