Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281999.1 | NP_001268928.1 | 16 | 1972 | ||
NM_013832.4 | NP_038860.2 | 17 | 1948 | |||
GenBank mRNA | AF086714.1 | - | 16 | 1820 | ||
AK156050.1 | - | 16 | 1936 | |||
BC005418.1 | - | 17 | 1962 | 3488024 |