Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013834.3 | NP_038862.2 | 3 | 1662 | ||
GenBank mRNA | AK081052.1 | - | 3 | 1255 | ||
AK088868.1 | - | 3 | 1665 | |||
BC094662.1 | - | 3 | 1648 | 30363219 | ||
U88566.1 | - | 3 | 1632 |
Gene Symbol: Serinc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC058550.1 | - | 628 |
Set Membership: |
G-protein coupled receptor |