Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015745.2 | NP_056560.1 | 4 | 3714 | ||
GenBank mRNA | BC017610.1 | - | 4 | 3714 | 4527589 | |
BC023069.1 | - | 4 | 1005 | |||
BC034121.1 | - | 4 | 833 | |||
BC035273.1 | - | 4 | 2146 | |||
BC040282.1 | - | 4 | 3698 | |||
BC042480.1 | - | 4 | 3676 | 5363924 | ||
BC042509.1 | - | 4 | 907 | |||
BC083133.1 | - | 4 | 3733 | 6492729 |