Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004798.3 | NP_004789.1 | 2 | 254 | ||
GenBank mRNA | AB002357.1 | - | 2 | 241 | ||
AK302403.1 | - | 2 | 249 | |||
BC136310.1 | - | 1 | 147 | 9020297 | ||
BC136311.1 | - | 1 | 147 | 9020298 | ||
DA805755.1 | - | 2 | 254 |