Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016670.3 | NP_057879.2 | 10 | 1117 | ||
GenBank mRNA | AF061270.1 | - | 10 | 1089 | ||
AK131788.1 | - | 9 | 1146 | |||
AK134000.1 | - | 11 | 1274 | |||
BC052701.1 | - | 9 | 1012 | 5721441 |