Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016676.5 | NP_057885.1 | 3 | 766 | ||
GenBank mRNA | AB232604.1 | - | 3 | 215 | ||
AF035646.1 | - | 3 | 456 | |||
AK008725.1 | - | 3 | 756 | |||
AK028320.1 | - | 3 | 737 | |||
AK087964.1 | - | 3 | 728 | |||
AK134988.1 | - | 3 | 686 | |||
AK146357.1 | - | 3 | 767 | |||
AK150319.1 | - | 3 | 726 | |||
AK150384.1 | - | 3 | 420 | |||
AK150791.1 | - | 3 | 716 | |||
AK151473.1 | - | 3 | 420 | |||
AK153315.1 | - | 3 | 726 | |||
AK153325.1 | - | 3 | 716 | |||
AK159383.1 | - | 3 | 731 | |||
AK159845.1 | - | 3 | 731 | |||
AK159988.1 | - | 3 | 725 | |||
AK160960.1 | - | 3 | 721 | |||
AK166010.1 | - | 3 | 737 | |||
AK168320.1 | - | 3 | 726 | |||
AK190935.1 | - | 2 | 68 | |||
AK193085.1 | - | 2 | 49 | |||
AK195457.1 | - | 3 | 202 | |||
AK202554.1 | - | 1 | 57 | |||
BC052735.1 | - | 716 | ||||
BC056374.1 | - | 3 | 717 | 6848840 | ||
CT010394.1 | - | 3 | 215 |