Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016858.2 | NP_058554.1 | 2 | 1437 | ||
GenBank mRNA | AB004665.1 | - | 2 | 1387 | ||
AK004974.1 | - | 2 | 1173 | |||
AK087967.1 | - | 2 | 1177 | |||
AK139821.1 | - | 2 | 1423 | |||
AK144706.1 | - | 2 | 1176 | |||
AK159674.1 | - | 2 | 1172 | |||
AK163783.1 | - | 2 | 1424 | |||
AK170483.1 | - | 2 | 1066 | |||
BC065076.1 | - | 2 | 1437 | 6815992 |