Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016907.3 | NP_058603.2 | 5, 6 | 1092 | ||
GenBank mRNA | AF099018.1 | - | 5, 6 | 1047 | ||
AK017342.1 | - | 4, 5 | 974 | |||
BC005769.1 | - | 4, 5 | 979 | 2650110 |